Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Siphon

Siphon, LDPE/PVC, 9 l/min, 1000 mm, 15 mm
  223-1513
 :   BURKLE
Siphon
  223-1513
 :  BURKLE
Siphon
Siphon

Siphon, LDPE/PVC, 9 l/min, 1000 mm, 15 mm

 
62,70 €
   
1 * 1 ST
 

Siphon pump in LDPE/PVC.

  • Good view of medium through the transparent tubing
  • With stop valve
  • Ideal for the occasional decanting of liquids

Easy to use model designed for the rapid decanting of aqueous liquids from elevated containers to those positioned lower down. Operate according to the siphon principle: Liquid needs only to be primed once. Not suitable for decanting strong acids and alkalis.

 
  • Flow rate:
    9 l/min
  • Description:
    Siphon
  • Material:
    LDPE/PVC
  • Immersion depth:
    1000 mm
  • Tube Ø:
    15 mm