Orders LinkContactUsLinkComponent
Repetitive pipette, Multipette® M4
Repetitive pipette, Multipette® M4
   Sustainable Options Available
Supplier:   EPPENDORF
Repetitive pipette, Multipette® M4
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Repetitive pipette, Multipette® M4
2 Options Available Below
Product Details & Documents
Manual dispenser, designed for maximum ease of use, precision and flexibility in terms of volume selection and replacement of associated Combitip advanced® tips.
  • Simple volume adjustment from 1 µl to 10 ml
  • Volume display with automatic recognition of the Combitip advanced® tip
  • Up to 100 dispensing cycles with one filling, integrated step counter
  • Capable of dispensing 112 different volumes
  • The Combitip® can be detached by applying slight pressure, avoiding any contact with and contamination of the tip
Combitip advanced® tips work on the direct displacement principle, ensuring that the correct volume is dispensed every time, regardless of the density of the liquid or its flow properties. This principle also ensures safer conditions when working with radioactive, aggressive, toxic substances or liquids with higher vapour pressures.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-2890
  • 613-2927
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001, ACT Label
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 613-2890
    Multipette® M4, incl. holder (for wall attachment or carousel stand)
    Multipette® M4, incl. holder (for wall attachment or carousel stand)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 440,00 €
    440,00 €
    1 * 1 ST
       440,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-2890
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 613-2927
    Multipette® M4 starter kit, incl. Combitip® rack, Combitip advanced® mixed pack
    Multipette® M4 starter kit, incl. Combitip® rack, Combitip advanced® mixed pack
    1 µl - 10 ml
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 467,00 €
    467,00 €
    1 * 1 KIT
       467,00 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    613-2927
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
Complementary Products
Product Information
  • Catalog #
    Packed
    Volume
    Colour code
    Sterile
    Availability
    Price
  • 613-2061
    100 pcs. (4 bags × 25 pcs.)
    1,0 ml
    Yellow
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2062
    100 pcs. (4 bags × 25 pcs.)
    2,5 ml
    Green
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2066
    100 pcs. (4 bags × 25 pcs.), each box containing one adapter
    50 ml
    Light grey
    -
    Log in for availability
    1 * 100 ST - 145,00 €
  • 613-2060
    100 pcs. (4 bags × 25 pcs.)
    0,5 ml
    Violet
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2063
    100 pcs. (4 bags × 25 pcs.)
    5,0 ml
    Blue
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2058
    100 pcs. (4 bags × 25 pcs.)
    0,1 ml
    White
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2059
    100 pcs. (4 bags × 25 pcs.)
    0,2 ml
    Light blue
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-2064
    100 pcs. (4 bags × 25 pcs.)
    10 ml
    Orange
    -
    Log in for availability
    1 * 100 ST - 112,00 €
  • 613-3857
    1 Combitips® advanced of each size (9 total) and Adapter advanced 25 & 50 ml
    0,1, 0,2, 0,5, 1,0, 2,5, 5,0, 10, 25, 50 ml
    Assorted
    -
    Log in for availability
    1 * 1 KIT - 16,40 €
  • 613-2065
    100 pcs. (4 bags × 25 pcs.), each box containing one adapter
    25 ml
    Red
    -
    Log in for availability
    1 * 100 ST - 132,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Charger carousel 2, for 6 Eppendorf Xplorer® or Xplorer® plus
    Charger carousel 2, for 6 Eppendorf Xplorer® or Xplorer® plus
    613-6971
    Log in for availability
    1 * 1 ST - 541,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette Holder 2, for one Eppendorf Multipette® M4, for Pipette Carousel 2 and Charger Carousel 2 or wall mounting, sticky tape included, without charging functionality
    Pipette Holder 2, for one Eppendorf Multipette® M4, for Pipette Carousel 2 and Charger Carousel 2 or wall mounting, sticky tape included, without charging functionality
    EPPE3116000.147
    Log in for availability
    1 * 1 ST - 45,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Velcro for pipette holder
    Velcro for pipette holder
    EPPE4982603.000
    Log in for availability
    1 * 1 ST - 6,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment cover
    Battery compartment cover
    EPPE4982601.008
    Log in for availability
    1 * 1 ST - 10,40 €