ContactUsLinkComponent
Nucleic acid building blocks
Nucleic acid building blocks
  761-0015
undefined
Nucleic acid building blocks
  761-0015
  • Description:
    Nucleic acid blocks
  • Pk:
    1
Coloured units representing phosphoric acids, purines and pyrimidines, for constructing DNA, t-RNA and RNA helices.
Useful for explaining replication and transcription.