Orders LinkContactUsLinkComponent
Standard ear
Standard ear
Ear in four parts, 340×160×190 mm
Catalog # 761-0004
Marketsource
Supplier:   3B Scientific
Standard ear
Catalog # 761-0004
Supplier:  3B Scientific
Marketsource
Standard ear
Standard ear
Ear in four parts, 340×160×190 mm
Price
288,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Representation of outer, middle and inner ear. Removable eardrum with hammer, anvil and stirrup as well as two part labyrinth with cochlea and auditory/balance nerve.
  • Three times life-size
  • On base
  • Four parts
Specifications
  • Description:
    Ear in four parts
  • W×D×H:
    340×160×190 mm
  • Magnification:
Recommendations