ContactUsLinkComponent
Polarising microscopes, BA310 POL
Polarising microscopes, BA310 POL
Supplier:  MOTIC
Certificates
undefined
Polarising microscopes, BA310 POL
Supplier:  MOTIC
Specifications
  • Catalog No:
  • MOTI1101001703971
  • MOTI1101001703941
  • MOTI1101001703951
  • MOTI1101001703961
  • 630-1822
  • MOTI1101001703981
  • Head:
  • 30° inclined
  • Eyepieces:
  • WF 10×/20 mm with pointer adjustment
  • Nosepiece:
  • Quadruple, reversed
  • Objectives:
  • Achromatic 4×, 10×, 40×, 60×
  • Condenser:
  • Abbe N.A. 0,90/1,25 with iris diaphragm
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Illumination:
  • Halogen
Specifications
Related Information
Camera Adapters
Product Information