Orders LinkContactUsLinkComponent
Microscope adapters for micromanipulators
Microscope adapters for micromanipulators
Supplier:   EPPENDORF
Microscope adapters for micromanipulators
Supplier:  EPPENDORF
Certificates
Microscope adapters for micromanipulators
Microscope adapters for micromanipulators
6 Options Available Below
Product Details & Documents
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.
Product Family Options
Product Information
  • Availability
    Price
  • 631-0217
    Nikon Eclipse® TS100
    Nikon Eclipse® TS100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 631-0213
    Zeiss Axiovert® 100/135
    Zeiss Axiovert® 100/135
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 631-0218
    Olympus® IX 50/70/71
    Olympus® IX 50/70/71
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 631-0214
    Leica® DMIRE 2 and DMIRB/E/HC
    Leica® DMIRE 2 and DMIRB/E/HC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 631-0224
    Zeiss Axiovert® 10/35
    Zeiss Axiovert® 10/35
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 732-6016
    Zeiss Axiovert 25/25 C40 C/40 CFL
    Zeiss Axiovert 25/25 C40 C/40 CFL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST