Orders LinkContactUsLinkComponent
Dispersing elements (aggregates) for POLYTRON® PT models
Dispersing elements (aggregates) for POLYTRON® PT models
Supplier:   KINEMATICA
Dispersing elements (aggregates) for POLYTRON® PT models
Supplier:  KINEMATICA
Certificates
Dispersing elements (aggregates) for POLYTRON® PT models
Dispersing elements (aggregates) for POLYTRON® PT models
10 Options Available Below
Product Details & Documents
Designation: EC - EasyClean version for the simplest cleaning or sterilisation (Standard)/2 - rotor/stator with 1 tooth row each.
Ordering information: Other dispersing aggregates available on request.
Product Family Options
Product Information
  • For unit
    Speed
    Stator/rotor Ø
    Length
    Volume
    Availability
    Price
  • 431-0142
    PT-DA 03/2 EC-E050
    PT-DA 03/2 EC-E050
    PT 1200 E / PT 2500 E
    3,45 m/s
    3,5/2,2 mm
    74 mm
    0,05 - 2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 530,00 €
    1 530,00 €
    1 * 1 ST
       1 530,00 €
  • 431-0051
    PT-DA 05/2 EC-E085
    PT-DA 05/2 EC-E085
    PT 1200 E / PT 2500 E
    3,9 m/s
    5,6/3,9 mm
    80 mm
    0,1 - 5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 620,00 €
    1 620,00 €
    1 * 1 ST
       1 620,00 €
  • 431-0054
    PT-DA 07/2 EC-E107
    PT-DA 07/2 EC-E107
    PT 2500 E /PT 1200 E
    6,5 m/s
    7,8/5 mm
    107 mm
    0,3 - 10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 700,00 €
    1 700,00 €
    1 * 1 ST
       1 700,00 €
  • 431-0053
    PT-DA 07/2SYN-E082
    PT-DA 07/2SYN-E082
    PT 1200E / PT 2500 E
    6,5 m/s
    7,8/5 mm
    82 mm
    0,3 - 10 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 837,00 €
    837,00 €
    1 * 25 ST
       837,00 €
  • 431-0058
    PT-DA 12/2 EC-E123
    PT-DA 12/2 EC-E123
    PT 1200 E / PT 2500 E
    11,8 m/s
    12/9 mm
    115 mm
    2 - 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 810,00 €
    1 810,00 €
    1 * 1 ST
       1 810,00 €
  • 431-0055
    PT-DA 12/2EC-E157
    PT-DA 12/2EC-E157
    PT 1200 E / PT 2500 E
    14,1 m/s
    12/9 mm
    154 mm
    2-250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 970,00 €
    1 970,00 €
    1 * 1 ST
       1 970,00 €
  • KINE11030299
    PT-DA 12/2EC-F154
    PT-DA 12/2EC-F154
    PT 10-35 GT/ PT 3100 D / PT 6100 D / PT 7100 D
    14,0 m/s
    — mm
    154 mm
    3 - 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 910,00 €
    1 910,00 €
    1 * 1 ST
       1 910,00 €
  • 431-0057
    PT-DA 12/2WEC-E157
    PT-DA 12/2WEC-E157
    PT 1200 E / PT 2500 E
    14,1 m/s
    12/9 mm
    154 mm
    2-250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 190,00 €
    2 190,00 €
    1 * 1 ST
       2 190,00 €
  • 431-0228
    PT-DA 25/2FEC-E192
    PT-DA 25/2FEC-E192
    PT 2500 E
    30,0 m/s
    25 /18 mm
    192 mm
    100 - 2500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 370,00 €
    2 370,00 €
    1 * 1 ST
       2 370,00 €
  • KINE11030302S
    PT-DA 20/2EC-F193S
    PT-DA 20/2EC-F193S
    PT 3100 D
    22 m/s
    20 mm
    193 mm
    20 - 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 120,00 €
    2 120,00 €
    1 * 1 ST
       2 120,00 €