Orders LinkContactUsLinkComponent
Burners
Burners
Supplier:   USBECK Laborgeräte
Burners
Certificates
Burners
8 Options Available Below
Product Details & Documents
DIN type.
  • Air regulation and pilot flame inside
Ordering information: Supplied without safety base, this needs to be ordered separately.
Product Family Options
Bunsen type
Product Information
  • For
    Availability
    Price
  • 451-1335
    Bunsen burner with needle valve
    Bunsen burner with needle valve
    Natural gas
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,80 €
    42,80 €
    1 * 1 ST
       42,80 €
  • 451-1325
    Bunsen burner with needle valve
    Bunsen burner with needle valve
    Propane
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,80 €
    42,80 €
    1 * 1 ST
       42,80 €
  • 451-1320
    Bunsen burner with stopcock
    Bunsen burner with stopcock
    Propane
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €
  • 451-1330
    Bunsen burner with stopcock
    Bunsen burner with stopcock
    Natural gas
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €
Teclu type
Product Information
  • For
    Availability
    Price
  • 451-1365
    Teclu burner with needle valve
    Teclu burner with needle valve
    Natural gas
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,60 €
    53,60 €
    1 * 1 ST
       53,60 €
  • 451-1355
    Teclu burner with needle valve
    Teclu burner with needle valve
    Propane
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,60 €
    53,60 €
    1 * 1 ST
       53,60 €
  • 451-1350
    Teclu burner with stopcock
    Teclu burner with stopcock
    Propane
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,60 €
    61,60 €
    1 * 1 ST
       61,60 €
  • 451-1360
    Teclu burner with stopcock
    Teclu burner with stopcock
    Natural gas
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,60 €
    61,60 €
    1 * 1 ST
       61,60 €
Safety base (provides extra stability for burners)
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Safety base for burners, provides extra stability and protects work surfaces
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,10 €
    7,10 €
    1 * 1 ST
       7,10 €