ContactUsLinkComponent
Gel documentation system, DigiDoc-It™ 130
  732-1111
undefined
Gel documentation system, DigiDoc-It™ 130
  732-1111
  • Description:
    DigiDoc-It® 130 system, without transilluminator
  • Pk:
    1
The DigiDoc-It® 130 imaging system connects to a computer (not included) and is designed as a simple and economical system for acquisition and documentation of gels and plates. The system combines a digital colour camera with megapixel resolution housed in the top of a compact, lightweight hood.
  • Gels can be viewed through the UV blocking viewpoint, which for maximum darkness can be closed when taking pictures
  • Exclusive capture software controls camera functions, image capture and image enhancement, annotation and archiving, and enables live preview of images
  • Software allows selection of user defined default settings and templates
  • Lightweight door opens for access to the UV transilluminator surface (transilluminator not supplied)
  • Handles allow hood to be easily removed from the transilluminator
Hood (W×D×H): 451×305×422 mm
 : Each system includes megapixel digital camera (DigiCam 130), hood enclosure, control software and ethidium bromide filter. An external PC is required but not supplied.