Air flow indicator
Air flow indicator
Catalog # 151-0187
Supplier: Dräger
Jump to:
Product Details & Documents
This air flow indicator for special applications can be used in all operations relating to draught detection.
Delivery information: Supplied with an aspirator bulb, ten air flow test tubes and two sealing caps.
Dokumente
DRAECH00216_TDS_EN
Technisches Datenblatt
Specifications
- Description:Air flow indicator
Accessories
Product Information
- Catalog #AvailabilityPrice
- Air flow test tubes