Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Systems
Gas Chromatographs
Accessories for Clarus® 500 GC
Accessories for Clarus® 500 GC
PERC09903392
:
PerkinElmer
Accessories for Clarus® 500 GC
PERC09903392
:
PerkinElmer
Description:
Replacement column nut, 1/16", connects capillary column to adapter
Seq:
07
Pk:
5