Order Entry
Austria
ContactUsLinkComponent
 
Vertical laminar flow workbench, Mini- V/PCR
Catalog # TELR1923477
Vertical laminar flow workbench, Mini- V/PCR
Catalog # TELR1923477

Specifications

  • Air velocity:
    0,4 m/s
  • Noise level:
    <58 dB(A)
  • Description:
    Mini-V/PCR
  • Model:
    Mini-V/PCR
  • Ext. W×D×H:
    670×520×1125 mm
  • Filters:
    "HEPA/ULPA H14 EN 1822: 99.999% at 0,3 μm particle size according to DOP
    Prefilter 85% ASHRAE"
  • Int. W×D×H:
    630×500×570 mm
  • Lighting:
    1600 lux
  • Weight:
    70 kg
  • Pk:
    1

Specifications

About this item

Vertical Laminar Flow Benches are an ideal solution for the handling of harmless substances and non hazardous biological samples where product sterility is required. They ensure class 100 area in accordance with US Fed. regulations. Std. 209 and BS 5195 for samples and product.

  • Reduced width and height for easy access to all laboratories
  • Foldable methacrylate UV resistant front panel
  • Stainless steel divided working tray (one piece in Mini V) to allow easy cleaning
  • LCD screen displays air speed, working hours and more

The small size Mini V is ideal for sample preparation for PCR. It is equipped with UV light as standard.

The unit is provided with one HEPA-H14 filter for the downflow, allowing operation in sterile and particle free conditions due to the continuous flushing of the working area by a unidirectional vertical and ultra filtered air flow, thus assuring full product protection.

Certifications: Class 4 according to ISO 14644-1; Grade A Annex 1 GMP.