ContactUsLinkComponent
Airflow detection powder, Draft Checker™, SP Bel-Art
Airflow detection powder, Draft Checker™, SP Bel-Art
Catalog # 215-6525
undefined
Airflow detection powder, Draft Checker™, SP Bel-Art
Catalog # 215-6525
Specifications
  • Description:
    Draft Checker™ 1,5 g
  • Pk:
    1
Specifications
About this item
An amorphous ultra-fine silica powder in a handy squeezy bottle.
  • Suspended powder permits visual estimation of airflow
  • Can be used for detecting leaks around windows, doors and ducts
Safety Note: Silica is inert and non-flammable but appropriate precautions should be taken to prevent ingestion or inhalation of the powder.