ContactUsLinkComponent
Universal upright freezers, GP models
Supplier:  Liebherr
Certificates
undefined
Universal upright freezers, GP models
Supplier:  Liebherr
Specifications
  • Catalog No:
  • LIEBGP1476
  • Model:
  • GP 1476
  • Cat. No.:
  • LIEBGP1476
  • Air circulation:
  • Non ventilated
  • Display:
  • LCD touch screen
  • Defrost:
  • Manual
  • Alarm:
  • Audible and visual
  • Shelves/drawers:
  • 4 plastic shelves
  • Door type:
  • 1 solid, reversible door, hinge right
  • Material:
  • Steel
  • Energy class:
  • A++
Specifications
Related Information
Product Information