Orders LinkContactUsLinkComponent
Electroporation plates
Electroporation plates
Supplier:   BTX
Electroporation plates
Supplier:  BTX
Certificates
Electroporation plates
Electroporation plates
4 Options Available Below
Product Details & Documents
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Product Family Options
Product Information
  • No. of wells
    Volume
    Availability
    Price
  • 732-0480
    Electroporation plates, 2 mm gap
    Electroporation plates, 2 mm gap
    25
    125 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
  • 732-0481
    Electroporation plates, 2 mm gap
    Electroporation plates, 2 mm gap
    25
    125 µl
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 605,00 €
    605,00 €
    1 * 6 ST
       605,00 €
  • 732-0476
    Electroporation plates, 2 mm gap
    Electroporation plates, 2 mm gap
    96
    125 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
  • 732-0477
    Electroporation plates, 4 mm gap
    Electroporation plates, 4 mm gap
    96
    250 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 320,00 €
    320,00 €
    1 * 1 ST
       320,00 €