FlashGel™ DNA system
About this item
FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.
- Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
- Gel runs can be monitored at the bench, in real time without UV light
- At least five times more sensitive than gels stained with ethidium bromide
Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel™ Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.
Delivery information: Supplied as a complete system (including FlashGel™ dock), separate components or as a starter kit (without FlashGel™ camera). The markers and dye are in a ready to load format, optimised for use with the system and recommended for best performance. The starter kit contains 9× FlashGel™ DNA cassettes (1,2%, 12+1 well); 1 ml FlashGel™ loading dye; and 150 µl FlashGel™ DNA marker (100 bp to 4 Kb).
Caution: Gel cannot be accessed. Unsuitable for recovery.
7 Options Available Below
Product Details & Documents
FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.
- Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
- Gel runs can be monitored at the bench, in real time without UV light
- At least five times more sensitive than gels stained with ethidium bromide
Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel™ Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.
Delivery information: Supplied as a complete system (including FlashGel™ dock), separate components or as a starter kit (without FlashGel™ camera). The markers and dye are in a ready to load format, optimised for use with the system and recommended for best performance. The starter kit contains 9× FlashGel™ DNA cassettes (1,2%, 12+1 well); 1 ml FlashGel™ loading dye; and 150 µl FlashGel™ DNA marker (100 bp to 4 Kb).
Caution: Gel cannot be accessed. Unsuitable for recovery.
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- LONZ57067
- 733-1610
- 733-1613
- 733-1611
- 733-1614
- 733-1615
- LONZ57051
- Model:
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- FlashGel™ DNA
- Gel size W×L:
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- 84×70 mm
- Separation range (bp):
- 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000 - 1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000
- Storage (cassettes):
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Room temperature for up to 5 months from manufacture
- Well volume:
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- 2 - 5 µl
- Cassette size W×D×H:
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- 107×115×17 mm
- Dock size W×D×H:
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
- 120×134×54 mm
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- LONZ57067FlashGel™ system, including FlashGel™ dock, FlashGel™ camera, 9× FlashGel™ DNA cassettes (1,2%, 12+1-well format), FlashGel™ loading dye and FlashGel™ DNA markerFlashGel™ system, including FlashGel™ dock, FlashGel™ camera, 9× FlashGel™ DNA cassettes (1,2%, 12+1-well format), FlashGel™ loading dye and FlashGel™ DNA markerLog in for availability1 * 1 ST2 720,00 €Specifications:
More
Specifications
Cat. No.LONZ57067ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1610FlashGel™ system for DNA, starter kit (without camera)Specifications:
More
Specifications
Cat. No.733-1610ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1613FlashGel™ DNA cassettes, 1,2% agarose, 16+1 double tierSpecifications:
More
Specifications
Cat. No.733-1613ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1611FlashGel™ DNA cassettes, 1,2% agarose, 12+1 single tierSpecifications:
More
Specifications
Cat. No.733-1611ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1614FlashGel™ DNA cassettes, 2,2% agarose, 12+1 single tierSpecifications:
More
Specifications
Cat. No.733-1614ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - 733-1615FlashGel™ DNA cassettes, 2,2% agarose, 16+1 double tierSpecifications:
More
Specifications
Cat. No.733-1615ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm - LONZ57051FlashGel™ recovery cassettes, 1,2% agarose, 8+1 double tierFlashGel™ recovery cassettes, 1,2% agarose, 8+1 double tierLog in for availability1 * 9 ST241,00 €Specifications:
More
Specifications
Cat. No.LONZ57051ModelFlashGel™ DNAGel size W×L84×70 mmSeparation range (bp)1,2% agarose: 50 - 4000
2,2% agarose: 10 - 1000Storage (cassettes)Room temperature for up to 5 months from manufactureWell volume2 - 5 µlCassette size W×D×H107×115×17 mmDock size W×D×H120×134×54 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Formaldehyde sample buffer
Product Information
- Catalog #AvailabilityPrice
- RNA marker (0,5 - 9 kb), 50 µL
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskFlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and maskLONZ57064Log in for availability1 * 1 KIT - 729,00 €
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettesFlashGel™ DNA marker (50 bp to 1 Kb), for 2,2% cassettes733-1616Log in for availability1 * 500 µl - 318,00 €
Product Information
- Catalog #AvailabilityPrice
- RNA marker (0,5 bp to 9 Kb)
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applicationsFlashGel™ DNA QuantLadder (100 bp to 1500 bp), 50 applications733-1621Log in for availability1 * 250 µl - 318,00 €
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettesFlashGel™ DNA marker (100 bp to 4 Kb), for 1.2% cassettes733-1620Log in for availability1 * 500 µl - 318,00 €
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettesFlashGel™ DNA marker (100 bp to 3 Kb), for 2 tier cassettes733-1622Log in for availability1 * 500 µl - 318,00 €
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)FlashGel™ loading dye, 5X, RNA native sample buffer (contains xylene cyanol)733-1619Log in for availability1 * 5 mL - 253,00 €
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ dock
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ recovery buffer, 2× 500 μl
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ power supply
Product Information
- Catalog #AvailabilityPrice
- FlashGel™ power supply pack, includes FlashGel™ dockFlashGel™ power supply pack, includes FlashGel™ dockLONZ57069Log in for availability1 * 1 KIT - 1 830,00 €