ContactUsLinkComponent
Extra-wide mini horizontal electrophoresis systems, Owl™ D3-14 and A6
Extra-wide mini horizontal electrophoresis systems, Owl™ D3-14 and A6
Catalog # 730-0109
undefined
Extra-wide mini horizontal electrophoresis systems, Owl™ D3-14 and A6
Catalog # 730-0109
Specifications
  • Model:
    Owl D3-14
  • Description:
    Extra-wide mini horizontal gel system
  • Buffer volume:
    800 ml
  • Gel size W×L:
    230×140 mm
  • Unit W×L×H:
    300×270×110 mm
  • Pk:
    1
Specifications
About this item
The Owl™ D3-14 and A6 wide gel systems offer a simple, convenient and rapid means of screening multiple samples on a single agarose gel. Ideal for screening PCR samples, plasmid preps, restriction mapping and cloning.
  • 25 to 200 samples can be run simultaneously on one gel, producing clear tight banding patterns with no ‘smiling'
  • Sample loading is greatly enhanced using microwell format combs engineered specifically for use with the system
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time when using microwell combs and a multichannel pipette, speeding up transfer and reducing the risk of sample order error
Delivery: Each system includes buffer chamber, SuperSafe lid with attached power leads, gasketed UVT gel tray, external caster and four combs (Owl D3-14: 50-microwell, 1,5 mm thick, Owl A6: 2×25- and 2×50-microwell, 1,5 mm thick).
Combs
Product Information