ContactUsLinkComponent
Large horizontal gel electrophoresis systems, Owl™ A2 and A5
Large horizontal gel electrophoresis systems, Owl™ A2 and A5
Certificates
undefined
Large horizontal gel electrophoresis systems, Owl™ A2 and A5
Certificates
Specifications
  • Catalog No:
  • 730-0154
  • 730-0103
  • Model:
  • Owl A2-BP
  • Owl A5
  • Gel size W×L:
  • 200×250 mm
  • 200×250 mm
  • Buffer volume:
  • 2300 ml
  • 2000 ml
  • Unit W×L×H:
  • 280×375×100 mm
  • 260×380×130 mm
Specifications
Related Information
Combs
Product Information
Frequently Bought Together