Order Entry
Austria
ContactUsLinkComponent
 
pH/°C meter, handheld, HI 991001
  662-4024
 :  Hanna
undefined
pH/°C meter, handheld, HI 991001
  662-4024
 :  Hanna

 

  • Temperature range:
    −5,0...+105,0 °C
  • Description:
    pH/°C meter HI 991001
  • Model:
    HI 991001
  • Temperature accuracy:
    ±0,5 °C (up to +60 °C) / ±1 (outside)
  • Temperature resolution:
    0,1 °C
  • Compliance:
    IP 67
  • Weight:
    0,210 kg
  • W×D×H:
    150×80×36 mm
  • pH accuracy:
    ±0,02
  • pH calibration:
    Automatic, 1 or 2 points with 2 sets of memorised buffer values (4,01 / 7,01 / 10,01 or 4,01 / 6,86 / 9,18)
  • pH range:
    −2,00...+16,00
  • pH resolution:
    0,01
  • Pk:
    1

 

 

HI 991001 meter offers IP 67 waterproof protection in a compact case.

  • Stainless steel bodied single probe measures pH and temperature
  • Ideal for harsh environments and heavy use
  • Large display with graphic symbols, showing simultaneous pH and temperature readings
  • Automatic pH calibration at 1 or 2 points using standard or NIST buffers
  • Long battery life (over 1500 hours of continuous use), remaining battery level display
 : Supplied complete with HI 1296D pH electrode with internal temperature sensor, pH 4 and pH 7 buffer sachets, electrode cleaning solution (2× 20 ml), batteries and rugged carrying case.