ContactUsLinkComponent
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Supplier:  WTW ONLINE
Certificates
undefined
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Supplier:  WTW ONLINE
Specifications
  • Catalog No:
  • 706-0524
  • 706-0525
  • Model:
  • FDO 700 IQ
  • FDO 700 IQ SW
  • Min. flow rate:
  • No flow required
  • No flow required
  • Response time:
  • <150 s
  • <60 s
  • DO concentration:
  • 0...20,00 mg/l
  • 0...20,00 mg/l
  • DO saturation:
  • 0...200,0%
  • 0...200,0%
  • DO barometric pressure range:
  • 10 bar
  • 10 bar
  • DO accuracy:
  • ±0,05 mg/l
  • ±0,05 mg/l
  • DO resolution:
  • 0,01 mg/l
  • 0,01 mg/l