ContactUsLinkComponent
Systematic On-Line analyser, TresCon® Uno
Systematic On-Line analyser, TresCon® Uno
Catalog # 708-0052
Supplier:  WTW ONLINE
undefined
Systematic On-Line analyser, TresCon® Uno
Catalog # 708-0052
Supplier:  WTW ONLINE
Specifications
  • Description:
    TresCon® Uno basic unit with nitrate module, TCU/N511
  • Pk:
    1
Specifications
About this item
The TresCon® Uno is a single channel analyzer designed for control and monitoring at waste water treatment plants.
  • Automatic diagnosis functions help the user with operation and maintenance
  • Modular design enables a quick and easy exchange or replacement of modules
  • Easy access and maintenance
  • Range of analyser modules are available for many parameters including ammonium-nitrogen, nitrate-nitrogen, orthophosphate, total phosphorus
  • With RS232 and RS485 interfaces
The compact size of TresCon® Uno at a reasonable price offers to the user a good price-performance-ratio for the measurement of nutrient parameters. The user interface within the system mounting is simple to operate and has a high resolution, backlit, graphics display.
Delivery: The TresCon® Uno consists of the basic controller unit, an analyser module, and a reagent tray. The system can be wall mounted.
For the full range of parameter modules and specification please contact the Avantor specialist.