ContactUsLinkComponent
Dissolved oxygen probe, StirrOx® G
Dissolved oxygen probe, StirrOx® G
Catalog # 664-0510
Supplier:  WTW
undefined
Dissolved oxygen probe, StirrOx® G
Catalog # 664-0510
Supplier:  WTW
Specifications
  • Type:
    StirrOx® G, self-strirring, incl. calibration vessel OxiCal®-ST
  • Description:
    Dissolved oxygen probe StirrOx® G
  • Pk:
    1
Specifications
About this item
Self-stirring oxygen probe for simultaneous stirring and measurement.
  • Single-handed operation for rapid series measurements
  • Constant approach flow for high reproducibility
  • Extremely low self-consumption of oxygen
  • Temperature compensation
  • Membrane leakage monitoring
Ordering information: Inclusive calibration vessel OxiCal®-ST.