Scrubbers for Kjeldahl mineralisation units
705-0488
: GERHARDT
- Description:Turbosog
- Pk:1
The Turbosog scubber unit condenses and neutralises aggressive acid vapours from the Kjeldahl digestion, including perchloric acid and other toxic gases.
- Pre-separator has two bottles for separation and washing
- Water consumption of 50 L/h
- Unit is easy to move