Order Entry
Austria
ContactUsLinkComponent
Liquid nitrogen storage vessels, TR
Liquid nitrogen storage vessels, TR
Supplier:  Air Liquide
Certificates
undefined
Liquid nitrogen storage vessels, TR
Supplier:  Air Liquide

Specifications

  • Catalog No:
  • 478-3115
  • 478-3114
  • 478-3117
  • 478-3116
  • 478-3113
  • 478-3112
  • Model:
  • TR 35
  • TR 26
  • TR 100
  • TR 60
  • TR 21
  • TR 11
  • Capacity:
  • 33,6 L
  • 26 L
  • 99 L
  • 60 L
  • 21,5 L
  • 12,2 L
  • Static holding time (days):
  • 140
  • 130
  • 180
  • 150
  • 119
  • 67
  • Daily evaporation:
  • 0,24 L
  • 0,2 L
  • 0,55 L
  • 0,4 L
  • 0,18 L
  • 0,18 L
  • Ø neck:
  • 50 mm
  • 50 mm
  • 50 mm
  • 50 mm
  • 50 mm
  • 50 mm
  • Ø×H:
  • 468×655 mm
  • 388×669 mm
  • 510×986 mm
  • 468×869 mm
  • 388×605 mm
  • 308×584 mm

Specifications

Frequently Bought Together