Orders LinkContactUsLinkComponent
Comparator lighting units
Comparator lighting units
Supplier:   Lovibond Tintometer
Comparator lighting units
Certificates
Comparator lighting units
Comparator lighting units
2 Options Available Below
Product Details & Documents
For consistent and accurate readings when using Lovibond comparator instruments, it is important to use a constant light source for matching the sample with the glasses.
Standardised lighting unit, Daylight 2000: This mains-operated benchtop unit is corrected to good Northern daylight using a tungsten halogen lamp and a calibrated colour temperature-correction filter. It guarantees uniform lighting conditions for colour matching, allowing consistent and accurate readings when using Lovibond test discs.
Portable lighting units: The portable lighting unit is a constant light source powered by standard batteries for colour matching in any location.
Product Family Options
Product Information
  • Availability
    Price
  • 705-0681
    Daylight 2000, lighting source for Comparator 2000+, mains-operated
    Daylight 2000, lighting source for Comparator 2000+, mains-operated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 090,00 €
    1 090,00 €
    1 * 1 ST
       1 090,00 €
  • 705-0682
    Daylight 2000 unit for Nessleriser, mains-operated
    Daylight 2000 unit for Nessleriser, mains-operated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €