Components for spectrophotometric colorimeters, PFXi-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
Certificates
Components for spectrophotometric colorimeters, PFXi-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
About this item
Frequently Bought Together
VWR®, Centrifuge Tubes, ASTM
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
VWR®, Twist-Seal Bags
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH electrodes, SenTix®
Supplier: WTW
Titration beakers
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Plastic cuvette for LICO
Supplier: Hach
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies