Orders LinkContactUsLinkComponent
Colony counters
Colony counters
Supplier:   IUL Instruments SA
Colony counters
Supplier:  IUL Instruments SA
Certificates
Colony counters
2 Options Available Below
Product Details & Documents
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.
Product Family Options
Product Information
  • Availability
    Price
  • IULA90008700
    DOT colony counter with 120 mm magnifying glass with light
    DOT colony counter with 120 mm magnifying glass with light
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 251,00 €
    1 251,00 €
    1 * 1 ST
       1 251,00 €
  • 710-0106
    DOT colony counter with 100 mm magnifying glass without light
    DOT colony counter with 100 mm magnifying glass without light
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 048,00 €
    1 048,00 €
    1 * 1 ST
       1 048,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 100 mm
    Centering Device 100 mm
    710-0111
    Log in for availability
    1 * 1 ST - 61,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 80 mm
    Centering Device 80 mm
    710-0110
    Log in for availability
    1 * 1 ST - 61,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 60 mm
    Centering Device 60 mm
    710-0109
    Log in for availability
    1 * 1 ST - 61,00 €