Orders LinkContactUsLinkComponent
Immersion thermostats
Immersion thermostats
Catalog #
Immersion thermostats
Catalog #
Supplier:  
Immersion thermostats
Immersion thermostats
Product Details & Documents
Heating immersion thermostats with wide range of functionality and features for your applications.
  • Mechanically adjustable pressure pump
  • Over-temperature and low liquid level protection to avoid any potential damage
  • Compact units suitable for use with virtually any tanks up to 50 litres and 26 mm wall thickness
  • Wetted parts and housing made of high quality stainless steel or plastic
  • External PT100 sensor connection
  • Integrated controller for 10 program steps for time and temperature dependent processes
Suitable for the most demanding applications; includes Intelligent Cascade Control, self optimising for optimum results and Temperature Control Features (TCF) for band limit, limit setting, co-speed factor and control dynamics. Temperature range up to 300 °C. Suitable for use with tanks up to 100 litres, bridge extends from 31 to 66 cm. Built-in cooling coil for counter cooling with tap water. Powerful electronically adjustable pressure and suction pump. 3 kW Heating capacity.
Max. flow rate/max. pressure:
SE-Z: 22 - 26 L/min, 700 mbar, 400 mbar suction