Temperature chart recorders, SC3
About this item
These recorders are used to record temperature simply and reliably. They provide the users with immediate access to data and suit applications in manufacture of pharmaceutical, medical and food products and vaccine storage.
- Withstand the rigours of transport and storage
- Accuracy and resolution conform to quality controls
- For recordings over 24 hours or seven days
W×D×H: 93×93×57 mm
Weight: 227 mm
Delivery information: Supplied with a single AA battery with a service life of two years and a red pen. Other diagrams are available on request.
4 Options Available Below
Product Details & Documents
These recorders are used to record temperature simply and reliably. They provide the users with immediate access to data and suit applications in manufacture of pharmaceutical, medical and food products and vaccine storage.
- Withstand the rigours of transport and storage
- Accuracy and resolution conform to quality controls
- For recordings over 24 hours or seven days
W×D×H: 93×93×57 mm
Weight: 227 mm
Delivery information: Supplied with a single AA battery with a service life of two years and a red pen. Other diagrams are available on request.
Product Family Options
Product Information
- RangeAccuracyResolutionW×D×HWeightAvailabilityPrice
- 620-0256Temperature chart recorder, SC377Temperature chart recorder, SC377–15…+10 °C±1,0 °C1,0 °C93×93×57 mm227 gLog in for availability1 * 1 ST405,00 €
- 620-0257Temperature chart recorder, SC386Temperature chart recorder, SC386–5…+20 °C±1,0 °C1,0 °C93×93×57 mm227 gLog in for availability1 * 1 ST478,00 €
- 620-0258Temperature chart recorder, SC387Temperature chart recorder, SC38710…35 °C±1,0 °C1,0 °C93×93×57 mm227 gLog in for availability1 * 1 ST405,00 €
- 620-0255Temperature chart recorder, SC367Temperature chart recorder, SC367–25…0 °C±1,0 °C1,0 °C93×93×57 mm227 gLog in for availability1 * 1 ST627,00 €
Charts
Product Information
- Catalog #DescriptionRecording timeRangeAvailabilityPrice
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- One-point NIST Traceable® calibration certificateOne-point NIST Traceable® calibration certificate620-1494Chart recordersLog in for availability1 * 1 ST - 97,50 €
Product Information
- Catalog #AvailabilityPrice
- Six red replacement pens