ContactUsLinkComponent
Accessories for thermoreactors Spectroquant® TR series
Catalog # 1.71203.0001
Supplier:  Merck
Accessories for thermoreactors Spectroquant® TR series
Catalog # 1.71203.0001
Supplier:  Merck
Specifications
  • Description:
    Thermosensor for TR420 and TR620
  • Pk:
    1
Specifications
About this item
The thermosensor measures the current temperature in the hole of the thermoreactor block and compares it with a specified temperature. The results can be transmitted to a PC for documentation purposes. Optimised, well matched system.