About this item
The XS and XA analytical balances provide fast and accurate weighing with flexibility. This is achieved with the introduction of SmartGrid technology, with the gridded weighing pan supported at the rear of the weighing chamber. Therefore the draught shield and drip tray can be completely, quickly and easily removed. The draught shield can be set to open the right hand door with the left slider and vice versa making loading more efficient. The design also features a large backlit touch screen that facilitates direct access to all functions and a standard range of applications, that include simple weighing, percentage weighing, piece counting and statistics. Software can be downloaded from the internet via RS232 connection.
- SmartGrid and ErgoClips for fast results and secure sample handling
- SmartScreen touch screen with SmartTrac graphical weighing-in guide on XA models
- FACT fully automatic time and temperature controlled internal adjustment and linearisation
- Adjustable and detachable display gives flexibility in situating the units
In addition to the benefits features listed previously the wider range of XS balances also feature:
More advanced applications including density determinations, formula weigh and totalise and calculate coefficients. GLP compliant documentation and settings can be password protected.
In addition to the benefits features listed previously the wider range of XS balances also feature: