pH electrode storage and cleaning solutions, Orion™
Solutions for storage and cleaning that prolong the working life of pH electrodes. Cleaning solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation.
- Cleaning solution A: for removing protein contaminants
- Cleaning solution B: for removing bacterial contaminants
- Cleaning solution C: for general cleaning
- Cleaning solution D: for removing oil and grease contaminants
Bottles include a small beaker to hold the solution and a plastic pipette to remove the old electrode filling solution.
Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples.
- Environmentally Preferable:
Frequently Bought Together