Redox electrodes, InLab® Redox
Supplier: Mettler - Toledo
Certificates
About this item
Redox electrodes with S7 connector.
- Ideal for testing the oxidation/reduction in factory and industrial waste (chromium and cyanide concentration), fish farming and aquariums (water quality), and swimming pools (sanitation levels)
- ARGENTHAL™ reference system with silver-ion trap (InLab® Redox Micro: without silver-ion trap)
- EQUITHAL® technology with symmetrical conducting layers guarantees shortest response times despite large changes in temperature
- Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
Temperature range: 0 to 100 °C
pH range: 0 - 14
Temperature range: 0...+100 °C
Ordering information: Supplied with test certificate and without cable. Other electrodes on request.
Frequently Bought Together