pH electrodes for Knick® Portamess® meters
Supplier: HAMILTON BONADUZ
Certificates
About this item
These electrodes are especially well suited for Knick® Portamess® equipment. The electrode head creates a hermetic seal with the Portamess® storage tube.
- Liq-Glass Knick Temp DIN: Everef reference system provides stable potentials resulting in long sensor lifetime
- Double Pore Knick/Polyplast Knick Temp DIN: Ag/AgCl reference system
- Specially formulated electrolytes allow pH measurements of aqueous and biological (protein containing) matrices
- Special diaphragm shapes for small sample volumes or semi-solid samples
pH range: 0 to 14
* Integrated temperature sensor
Ordering information: Supplied with individual certificate and serial number.
Frequently Bought Together