Centrefeed rolls and wipes, Tork
Supplier: Essity
Certificates
About this item
Centrefeed rolls and wipes for Tork dispenser systems available in various qualities.
- Tork Heavy-Duty: Soft premium quality for heaviest wiping tasks - with a better, faster absorption than other towels and wipes
- Tork Plus: Improved performance tissues and wipes that meet or exceed basic requirements - with a good absorption, brightness and strength
- Tork Basic: Standard quality tissues and wipes for basic demands
Frequently Bought Together