ContactUsLinkComponent
Wipe rolls, Multisoft® Air
Wipe rolls, Multisoft® Air
Catalog # 115-2728
undefined
Wipe rolls, Multisoft® Air
Catalog # 115-2728
Specifications
  • Packed:
    1 roll with 500 sheets
  • L×W:
    400×250 mm
  • Environmentally Preferable:
  • Pk:
    1 Roll
Specifications
About this item
High quality wipes for cleaning and polishing sensitive surfaces such as metal, plastics, wood, or finishes. Multi-soft cotton fleece roll with 500 sheets.
  • Extremely soft, highly absorbent and solvent-resistant
  • Strong, low-linting with special embossment
Colour: White
Colour: White