Micro-Ubbelohde viscometers
Supplier: SI Analytics
Certificates
About this item
Viscometers, with suspended ball level, for determination of relative or absolute kinematic viscosity.
- Suitable for small samples and short running times
- Ring marks for checking by manually
- Uncalibrated or calibrated with constant for automatic measurements
Certifications: DIN 51 562-2
Frequently Bought Together