Orders LinkContactUsLinkComponent
WHEATON® Scintillation Vials
WHEATON® Scintillation Vials
Supplier:  DWK Life Sciences
Certificates
WHEATON® Scintillation Vials
WHEATON® Scintillation Vials
15 Options Available Below
Product Details & Documents
These vials are made from WHEATON® 180 low potassium borosilicate glass. The background counts of these vials are consistently low, the ultraviolet transmission is high.
  • Available with attached or unattached screw caps
  • Utility trays can be used to store samples
 : Glass conforms to ASTM E438 Type I, Class A.
 : Packed in five utility trays with each partition tray holding 100 vials.
Product Family Options
Caps attached to vials
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap Size
    Cap material
    Septum
    Availability
    Price
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 448,00 €
    448,00 €
    1 * 500 ST
       448,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 453,00 €
    453,00 €
    1 * 500 ST
       453,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 488,00 €
    488,00 €
    1 * 500 ST
       488,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 571,00 €
    571,00 €
    1 * 500 ST
       571,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    Urea
    PE disc
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 552,00 €
    552,00 €
    1 * 500 ST
       552,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 535,00 €
    535,00 €
    1 * 500 ST
       535,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 540,00 €
    540,00 €
    1 * 500 ST
       540,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 603,00 €
    603,00 €
    1 * 500 ST
       603,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    Urea
    PE disc
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 631,00 €
    631,00 €
    1 * 500 ST
       631,00 €
Caps packaged separately
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap Size
    Cap material
    Septum
    Availability
    Price
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 453,00 €
    453,00 €
    1 * 500 ST
       453,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 468,00 €
    468,00 €
    1 * 500 ST
       468,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 502,00 €
    502,00 €
    1 * 500 ST
       502,00 €
  • 20,00 ml
    28×61 mm
    Yes
    22 mm
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 586,00 €
    586,00 €
    1 * 500 ST
       586,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 535,00 €
    535,00 €
    1 * 500 ST
       535,00 €
  • 20,00 ml
    28×61 mm
    Yes
    24 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 540,00 €
    540,00 €
    1 * 500 ST
       540,00 €