Orders LinkContactUsLinkComponent
Scintillation vials, PET, WHEATON®
Supplier:   DWK Life Sciences
Scintillation vials, PET, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Scintillation vials, PET, WHEATON®
Scintillation vials, PET, WHEATON®
15 Options Available Below
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
  • Clarity of glass
  • Can be safely incinerated
  • Lightweight and virtually unbreakable
* With cap on
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap material
    Septum
    Availability
    Price
  • 20,00 ml
    27×61 mm
    Yes
    PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 394,00 €
    394,00 €
    1 * 500 ST
       394,00 €
  • 20,00 ml
    27×61* mm
    Yes
    PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 394,00 €
    394,00 €
    1 * 500 ST
       394,00 €
  • 20,00 ml
    27×61* mm
    Yes
    Urea
    Poly-seal cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 576,00 €
    576,00 €
    1 * 500 ST
       576,00 €
  • 20,00 ml
    27×61 mm
    Yes
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 658,00 €
    658,00 €
    1 * 1.000 ST
       658,00 €
  • 20,00 ml
    27×61 mm
    Yes
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 727,00 €
    727,00 €
    1 * 1.000 ST
       727,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 776,00 €
    776,00 €
    1 * 1.000 ST
       776,00 €
  • 20,00 ml
    27×61 mm
    Yes
    PE
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 707,00 €
    707,00 €
    1 * 1.000 ST
       707,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 943,00 €
    943,00 €
    1 * 1.000 ST
       943,00 €
  • 20,00 ml
    27×61 mm
    Yes
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 394,00 €
    394,00 €
    1 * 500 ST
       394,00 €
  • 20,00 ml
    27×61 mm
    Yes
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 394,00 €
    394,00 €
    1 * 500 ST
       394,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 576,00 €
    576,00 €
    1 * 500 ST
       576,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 438,00 €
    438,00 €
    1 * 500 ST
       438,00 €
  • 20,00 ml
    27×61* mm
    Yes
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 394,00 €
    394,00 €
    1 * 500 ST
       394,00 €
  • 20,00 ml
    27×61* mm
    Yes
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 389,00 €
    389,00 €
    1 * 500 ST
       389,00 €
  • 20,00 ml
    27×61* mm
    Yes
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 438,00 €
    438,00 €
    1 * 500 ST
       438,00 €