Closures, PPCO, natural, with colour coder, 11 mm
Supplier: Thermo Fisher Scientific
Certificates
About this item
Identify samples quickly with high profile closures with colour coders for micro packaging vials. Polypropylene polymer closures are ready to use and have no O-ring to fall out or contaminate contents.
- RNase/DNase free (only sterile closures)
- Permanently attached, colour coded inserts are free of toxic heavy metals
- Inserts do not come in contact with vial contents
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Closures are bulk packed without vials, 500 per inner bag, 1000 per case
Specifications
- Catalog No:
- 215-1648
- 215-1650
- 215-1649
- 215-1630
- 215-1188
- 215-1632
- 215-1631
- 215-1191
- 215-1638
- 215-1527
- 215-1263
- 215-1633
- 215-1526
- 215-1529
- 215-1190
- 215-1640
- 215-1528
- 215-1189
- 215-1639
- 215-1531
- 215-1530
- 215-1544
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement