Orders LinkContactUsLinkComponent
Sample bottles without push-on Closure
Supplier:   GRAVIS
Sample bottles without push-on Closure
Supplier:  GRAVIS
Certificates
Sample bottles without push-on Closure
Sample bottles without push-on Closure
6 Options Available Below
Product Details & Documents
Chemistry glass, white or amber, without push-on cap.
  • Amber glass to protect light-sensitive materials
 : Push-on PE caps are available separately.
Product Family Options
Sample Bottles, Clear Glass, without Cap
Product Information
  • Capacity
    Height
    Neck-Ø
    Availability
    Price
  • 31 ml
    62,2 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 128,00 €
    128,00 €
    1 * 210 ST
       128,00 €
  • 67 ml
    76 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 140 ST
    1 * 140 ST - 111,00 €
    111,00 €
    1 * 140 ST
       111,00 €
  • 96,5 ml
    75,7 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 86 ST
    1 * 86 ST - 93,90 €
    93,90 €
    1 * 86 ST
       93,90 €
  • 11,5 ml
    50 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 220 ST
    1 * 220 ST - 132,00 €
    132,00 €
    1 * 220 ST
       132,00 €
Sample Bottles, Amber Glass, without Cap
Product Information
  • Capacity
    Height
    Neck-Ø
    Availability
    Price
  • 18 ml
    38,5 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 122,00 €
    122,00 €
    1 * 210 ST
       122,00 €
  • 25 ml
    51 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 171,00 €
    171,00 €
    1 * 210 ST
       171,00 €
Push-on Caps for Sample Bottles
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Natural PE cap, Ø 30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,50 €
    53,50 €
    1 * 1.000 ST
       53,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Natural PE cap, Ø 22 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 47,50 €
    47,50 €
    1 * 1.000 ST
       47,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Natural PE cap, Ø 40 mm
    Log in for availability
    Log in for availability
    1 * 86 ST
    1 * 86 ST - 26,80 €
    26,80 €
    1 * 86 ST
       26,80 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Natural PE cap, Ø 30 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 6,90 €
    6,90 €
    1 * 100 ST
       6,90 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Natural PE cap, Ø 22 mm
    Log in for availability
    Log in for availability
    1 * 220 ST
    1 * 220 ST - 21,10 €
    21,10 €
    1 * 220 ST
       21,10 €