Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
Certificates
About this item
For storage in the freezer or in liquid nitrogen vapor. Suitable for temperatures from −196 to +121 °C.
- Certified non-cytotoxic and free of detectable endotoxins, DNase/RNase and human DNA
- With writing area and graduation
PP, with coloured cap, sterile (SAL 10⁻³).
* without writing area
Specifications
- Catalog No:
- 479-4153
- 479-0193
- 479-0196
- 479-4155
- 479-4152
- 479-4151
- 479-4150
- 479-4154
- 479-0195
- 479-0192
- 479-0197
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)