VWR® ND8, Crimp Neck Vials
Supplier: VWR Collection
Certificates
About this item
These vials are for use on instruments from the following manufacturers: Agilent, Beckman, Carlo Erba, CTC, Fisons, PerkinElmer, Shimadzu, Thermo Scientific, VWR®/Hitachi, more.
- Flat, round or conical bottom
- In clear or amber glass
- For almost all autosamplers
Broad selection of crimp neck vials ND8 available: Crimp neck vials and micro-vials ND8 can be closed with 8 mm aluminium caps, 9 mm PE caps or with 8 mm push-on caps. However, micro-vials often need an adapter to run in the autosampler. They often have a conical bottom shape, so need an adapter to stand.
Related Information
Aluminium crimp caps, ND8 with central hole, 4 mm and septum
Product Information
Frequently Bought Together