Seals and closures for 8 mm screw vials, Chromacol™
Supplier: CHROMACOL
Certificates
About this item
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetic acid glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Laboratory trays, low or high profile
Supplier: Gilac PROFESSIONNEL
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, Myristicin
Supplier: Merck
Seals and closures for 8 mm screw vials, Chromacol™
Supplier: CHROMACOL
Membrane filters with grid
Supplier: Sartorius