Orders LinkContactUsLinkComponent
Non-return valves
Non-return valves
Supplier:   KARTELL
Non-return valves
Supplier:  KARTELL
Certificates
Non-return valves
Non-return valves
2 Options Available Below
Product Details & Documents
PP, translucent. Prevent pressure loss and backflow. Maximum working pressure 2 bar and minimum working pressure  0,07 bar at room temperature (20 °C).
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Tapered ends to ensure that tubes are held firmly
Product Family Options
Product Information
  • Tubing size
    Availability
    Price
  • 7,6 - 10 mm ID
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 37,50 €
    37,50 €
    1 * 10 ST
       37,50 €
  • 9,2 - 12,5 mm ID
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 69,00 €
    69,00 €
    1 * 10 ST
       69,00 €