Cutter for polymeric tubing, clean-cut
Catalog # 554-1106
Supplier: VICI JOUR RESEARCH
Specifications
- Description:Cutter
- Pk:1
Specifications
About this item
This cutter provides burr-free perpendicular cuts of tubing up to ¹/₈" in PEEK, PTFE, ETFE and other polymeric tubing.
- Safety lock secures the blade
- No distortion of outside or closing of inside of the tubing