Order Entry
Austria
Orders LinkContactUsLinkComponent
Drying tubes, straight

Drying tubes, straight

130 mm, 13 mm
Catalog # 201-9110
Supplier:   Lenz Laborglas
Drying tubes, straight
Catalog # 201-9110
Supplier:  Lenz Laborglas
Drying tubes, straight
Drying tubes, straight

130 mm, 13 mm

Price
8,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, clear. One bulb and glass tubing connection.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Documents

Specifications
  • Ø int.:
    13 mm
  • Length:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.