Drying tubes with standard ground joints, U-shaped
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors with 6 mm external diameter.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Frequently Bought Together