Drying tubes with standard ground joints, angled
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Clear DURAN® borosilicate 3.3 glass. Angled at 75° with ground joint cone.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Frequently Bought Together