Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
Supplier:   LUDI
Caps, LABOCAP, without grip
Supplier:  LUDI
Certificates
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
24 Options Available Below
Product Details & Documents
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Product Family Options
Product Information
  • For Ø ext.
    Colour
    Availability
    Price
  • 9-10 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 21,20 €
    21,20 €
    1 * 10 ST
       21,20 €
  • 12 - 13 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 18,10 €
    18,10 €
    1 * 10 ST
       18,10 €
  • 12 - 13 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 18,10 €
    18,10 €
    1 * 10 ST
       18,10 €
  • 12 - 13 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 18,10 €
    18,10 €
    1 * 10 ST
       18,10 €
  • 14-15 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,40 €
    24,40 €
    1 * 10 ST
       24,40 €
  • 14 - 15 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,40 €
    24,40 €
    1 * 10 ST
       24,40 €
  • 15 - 16 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 19,40 €
    19,40 €
    1 * 10 ST
       19,40 €
  • 15-16 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 19,40 €
    19,40 €
    1 * 10 ST
       19,40 €
  • 15-16 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 19,40 €
    19,40 €
    1 * 10 ST
       19,40 €
  • 15-16 mm
    Green
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 19,30 €
    19,30 €
    1 * 10 ST
       19,30 €
  • 17 - 18 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,10 €
    20,10 €
    1 * 10 ST
       20,10 €
  • 17 - 18 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,10 €
    20,10 €
    1 * 10 ST
       20,10 €
  • 17 - 18 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,10 €
    20,10 €
    1 * 10 ST
       20,10 €
  • 17-18 mm
    Yellow
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 21,20 €
    21,20 €
    1 * 10 ST
       21,20 €
  • 19 - 20 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 21,20 €
    21,20 €
    1 * 10 ST
       21,20 €
  • 19-20 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 21,20 €
    21,20 €
    1 * 10 ST
       21,20 €
  • 19-20 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 20,60 €
    20,60 €
    1 * 10 ST
       20,60 €
  • 21-23 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,60 €
    36,60 €
    1 * 10 ST
       36,60 €
  • 21-23 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,60 €
    36,60 €
    1 * 10 ST
       36,60 €
  • 24-26 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,20 €
    40,20 €
    1 * 10 ST
       40,20 €
  • 24-26 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,20 €
    40,20 €
    1 * 10 ST
       40,20 €
  • 24-26 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,20 €
    40,20 €
    1 * 10 ST
       40,20 €
  • 28-30 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 42,00 €
    42,00 €
    1 * 10 ST
       42,00 €
  • 28-30 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 42,00 €
    42,00 €
    1 * 10 ST
       42,00 €